
1,3-Bis(1H,1H,2H,2H-tridecafluoroooct-1-yl)-1,1,3,3-tetramethyldisiloxane
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71363-70-7
:
Formula:C₂₀H₂₀F₂₆OSi₂
MDL Number:MFCD00270970
:826.502 g/mol
Boiling Pt:81…82 °C (11 mm Hg)
Density:1.46 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together