
1,1'-(1,1,2,2-Tetrafluoroethane-1,2-diyl)bis(4-fluorobenzene) 98%
Catalog # APOSPC2215-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:290.204 g/mol
Melting Pt:96…97 °C
:GHS07
Storage Temperature:Ambient
CAS Number:4100-99-6
Formula:C₁₄H₈F₆
MDL Number:MFCD01631642