Order Entry
Czech Republic
ContactUsLinkComponent
(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
Catalog # APOSPC210052-1G
Supplier:  Apollo Scientific
CAS Number:  
(Z)-2-[2-(4-Fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethenimidamide
Catalog # APOSPC210052-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.276 g/mol
Storage Temperature:Ambient
CAS Number:1255791-17-3
Formula:C₁₄H₁₂FN₅O₂
MDL Number:MFCD17780265