2-(Dimethylamino)-4-fluoro-3-formylbenzonitrile
Catalog # APOSPC210041-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:192.19 g/mol
Storage Temperature:Ambient
CAS Number:1515395-53-5
Formula:C₁₀H₉FN₂O
MDL Number:MFCD28101537