N-(4-Bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide
Catalog # APOSPC210037-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:298.057 g/mol
Storage Temperature:Ambient
CAS Number:870838-52-1
Formula:C₉H₇BrF₃NO₂
MDL Number:MFCD28101534