(3-Chlorophenyl)trifluoroboranuide tetrabutylazanium
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:411206-87-6
:
Formula:C₆H₄BClF₃·N(C₄H₉)₄
MDL Number:MFCD28101523
:421.819 g/mol
Storage Temperature:Ambient
Frequently Bought Together