6-Chloro-5-(trifluoromethyl)pyridin-3-amine 95+%
Catalog # APOSPC201388-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:196.558 g/mol
Storage Temperature:Ambient
CAS Number:99368-68-0
Formula:C₆H₄ClF₃N₂
MDL Number:MFCD11101042