(R)-1-(4-(Trifluoromethyl)phenyl)ethanamine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:856645-99-3
:
Formula:C₉H₁₀F₃N·ClH
MDL Number:MFCD11100948
:225.639 g/mol
Storage Temperature:Ambient
Frequently Bought Together