Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Amino-3-fluorophenyl)ethanone
1-(4-Amino-3-fluorophenyl)ethanone 98%
1-(4-Amino-3-fluorophenyl)ethanone 98%
1-(4-Amino-3-fluorophenyl)ethanone 98%
APOSPC201298-5G
:
Apollo Scientific
:
1-(4-Amino-3-fluorophenyl)ethanone 98%
APOSPC201298-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
153.154 g/mol
:
Ambient
:
73792-22-0
:
C₈H₈FNO
:
MFCD12032124