(S)-2-Amino-2-(2-fluorophenyl)ethanol hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1269773-22-9
:
Formula:C₈H₁₀FNO·ClH
MDL Number:MFCD12758111
:191.63 g/mol
Storage Temperature:Ambient
Frequently Bought Together