
2-Ethoxy-6-(trifluoromethyl)pyridine 95+%
Catalog # APOSPC201090-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.15 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1245563-18-1
Formula:C₈H₈F₃NO
MDL Number:MFCD17015840