1-(2-(Trifluoromethyl)phenyl)cyclopropanamine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1228879-24-0
:
Formula:C₁₀H₁₀F₃N·ClH
MDL Number:MFCD09028131
:237.649 g/mol
Storage Temperature:Ambient
Frequently Bought Together