(5-Fluoropyridin-2-yl)methanamine hydrochloride 95+%
Catalog # APOSPC201084-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:162.593 g/mol
Storage Temperature:Ambient
CAS Number:1228788-32-6
Formula:C₆H₇FN₂·ClH
MDL Number:MFCD11505886