
1-(Difluoromethyl)-4-iodo-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1041205-43-9
:
Formula:C₄H₃F₂IN₂
MDL Number:MFCD18633160
:243.981 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together