0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazol-5-amine
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazol-5-amine
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazol-5-amine
APOSPC200599-1G
:
Apollo Scientific
:
3-[4-(Trifluoromethyl)phenyl]-1H-pyrazol-5-amine
APOSPC200599-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
227.186 g/mol
:
153…155 °C
:
GHS07
:
Ambient
:
130599-34-7
:
C₁₀H₈F₃N₃
:
MFCD06738295