6,8-Difluoro-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:228728-19-6
:
Formula:C₁₀H₅F₂NO₃
MDL Number:MFCD00973973
:225.148 g/mol
Storage Temperature:Ambient
Frequently Bought Together