Methyl 2-[(4-bromo-2-chlorophenyl)amino]-3,4-difluoro-5-nitrobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1582770-01-1
:
Formula:C₁₄H₈BrClF₂N₂O₄
MDL Number:MFCD27980618
:421.578 g/mol
Storage Temperature:Ambient
Frequently Bought Together