Potassium (5-bromo-2-methoxyphenyl)trifluoroboranuide
Catalog # APOSPC200454-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:292.93 g/mol
Storage Temperature:Ambient
Formula:C₇H₆BBrF₃O·K
MDL Number:MFCD27980605