Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
Catalog # APOSPC200452-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-{[3-(Trifluoromethyl)phenyl]amino}-1,3-thiazol-5-yl)ethan-1-one
Catalog # APOSPC200452-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.273 g/mol
Melting Pt:193…195 °C
Storage Temperature:Ambient
CAS Number:1582770-06-6
Formula:C₁₂H₉F₃N₂OS
MDL Number:MFCD27980587