
2-Chloro-6-(trifluoromethoxy)pyridine
Catalog # APOSPC200446-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:81…83 °C (26 mm Hg)
:197.542 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1221171-70-5
Formula:C₆H₃ClF₃NO
MDL Number:MFCD18909427