Tetrabutylazanium trifluoro({3-[(4-methylpiperazin-1-yl)methyl]phenyl})boranuide
Catalog # APOSPC200441-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:499.547 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₇BF₃N₂·N(C₄H₉)₄
MDL Number:MFCD27957317