Order Entry
Czech Republic
ContactUsLinkComponent
1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one
1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one
Catalog # APOSPC200433-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(4-Fluorophenyl)-4-methyl-1,3-oxazol-5-yl]ethan-1-one
Catalog # APOSPC200433-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :219.212 g/mol
Melting Pt:90…92 °C
Storage Temperature:Ambient
CAS Number:1565845-75-1
Formula:C₁₂H₁₀FNO₂
MDL Number:MFCD27957301