N-(3,5-Difluorophenyl)-2,2,2-trifluoroacetamide
Catalog # APOSPC200428-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.115 g/mol
Storage Temperature:Ambient
CAS Number:1378573-08-0
Formula:C₈H₄F₅NO
MDL Number:MFCD20546924