6-Chloro-N-[3-(trifluoromethyl)phenyl]pyrimidin-4-amine
Catalog # APOSPC200391-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:273.642 g/mol
Storage Temperature:Ambient
CAS Number:872510-82-2
Formula:C₁₁H₇ClF₃N₃
MDL Number:MFCD10000669