Methyl 1-(4-fluorophenyl)-4-oxocyclohexane-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:80912-51-2
:
Formula:C₁₄H₁₅FO₃
MDL Number:MFCD22423113
:250.266 g/mol
Storage Temperature:Ambient
Frequently Bought Together