Order Entry
Czech Republic
ContactUsLinkComponent
2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one
2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one
Catalog # APOSPC200345-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(Trifluoromethyl)phenyl]-3,4-dihydropyrimidin-4-one
Catalog # APOSPC200345-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :240.181 g/mol
Melting Pt:185…187 °C
Storage Temperature:Ambient
CAS Number:125903-84-6
Formula:C₁₁H₇F₃N₂O
MDL Number:MFCD12107651