
Potassium trifluoro(3-phenyl-1H-pyrazol-5-yl)borate
Catalog # APOSPC200109-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
Melting Pt:180 °C
:250.07 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1402242-81-2
Formula:C₉H₇BF₃N₂·K
MDL Number:MFCD25948965