Order Entry
Czech Republic
ContactUsLinkComponent
1-(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)ethanol
1-(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)ethanol
Catalog # APOSPC200104-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl)ethanol
Catalog # APOSPC200104-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :194.154 g/mol
Storage Temperature:Ambient
Formula:C₇H₉F₃N₂O
MDL Number:MFCD25901665