4,6-Dioxo-9-(trifluoromethyl)-2,4,5,6-tetrahydro-1H-benzo[g]imidazo[1,2,3-ij]pteridin-12-ium chloride
Catalog # APOSPC200098-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:344.676 g/mol
Storage Temperature:Ambient
CAS Number:353245-38-2
Formula:C₁₃H₈F₃N₄O₂·Cl
MDL Number:MFCD25948722