6-Fluoro-3-iodoimidazo[1,2-a]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1219127-01-1
:
Formula:C₇H₄FIN₂
MDL Number:MFCD20483025
:262.023 g/mol
Melting Pt:178…181 °C
Storage Temperature:Ambient
Frequently Bought Together