5-[(2-Fluoro-4-iodophenyl)amino]-6-(methoxycarbonyl)-1-methyl-3-(prop-2-en-1-yl)-1,3-benzodiazol-1-ium iodide
Supplier: Apollo Scientific
Certificates
About this item
:
Formula:C₁₉H₁₈FIN₃O₂·I
MDL Number:MFCD22689148
:593.172 g/mol
Melting Pt:202 °C
Storage Temperature:Ambient
Frequently Bought Together