
(2,2,3,3-Tetrafluoro-1,4-benzodioxan-6-yl)ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:540738-37-2
:
Formula:C₁₀H₆F₄O₃
MDL Number:MFCD08275274
:250.146 g/mol
Boiling Pt:97 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together