Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-6-(trifluoromethyl)benzimidazole-2-thiol 97%
Irritant
4-Bromo-6-(trifluoromethyl)benzimidazole-2-thiol 97%
Catalog # APOSPC1595-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Bromo-6-(trifluoromethyl)benzimidazole-2-thiol 97%
Catalog # APOSPC1595-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :297.095 g/mol
Boiling Pt:>300 °C
Melting Pt:>300 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:175135-17-8
Formula:C₈H₄BrF₃N₂S
MDL Number:MFCD00067737