
2,3-Difluoro-6-(trifluoromethyl)benzonitrile 95+%
Catalog # APOSPC1513-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.1 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:186517-05-5
Formula:C₈H₂F₅N
MDL Number:MFCD06658270