
3'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:408367-99-7
:
Formula:C₁₄H₉F₃O₂
MDL Number:MFCD04039234
:266.215 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together