

4'-(Trifluoromethyl)[1,1'-biphenyl]-2-carboxaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:84392-23-4
:
Formula:C₁₄H₉F₃O
MDL Number:MFCD01631855
:250.216 g/mol
Density:1.27 g/cm³ (25 °C)
UN:3082
ADR:9+EHS,III
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together