
5-(3-Fluorophenyl)-2-furoic acid
Catalog # APOSPC11152-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:206.17 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:54022-97-8
Formula:C₁₁H₇FO₃
MDL Number:MFCD04039229