Order Entry
Czech Republic
ContactUsLinkComponent
3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole
3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole
Catalog # APOSPC110032-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[4-(2,4-Difluorophenoxy)phenyl]-1H-pyrazole
Catalog # APOSPC110032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :272.25 g/mol
Storage Temperature:Ambient
CAS Number:861213-56-1
Formula:C₁₅H₁₀F₂N₂O
MDL Number:MFCD06409160