
5-Amino-2-(4-bromophenoxy)benzotrifluoride
Catalog # APOSPC10712-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.116 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:74338-20-8
Formula:C₁₃H₉BrF₃NO
MDL Number:MFCD16987557