Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(4-Fluorophenoxy)benzyl bromide
4-(4-Fluorophenoxy)benzyl bromide
4-(4-Fluorophenoxy)benzyl bromide
APOSPC10711-1G
:
Apollo Scientific
:
4-(4-Fluorophenoxy)benzyl bromide
APOSPC10711-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
281.12 g/mol
:
GHS05
:
Ambient
:
83642-03-9
:
C₁₃H₁₀BrFO
:
MFCD15146807