
2-Iodo-5-(trifluoromethoxy)toluene
Catalog # APOSPC1061-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:302.032 g/mol
Density:1.776 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:886763-05-9
Formula:C₈H₆F₃IO
MDL Number:MFCD07368714