
2-Fluoro-6-nitrobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:887266-98-0
:
Formula:C₆H₃ClFNO₄S
MDL Number:MFCD07368711
:239.609 g/mol
Melting Pt:91 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together