2-(4-Chloro-3-fluorophenoxy)benzaldehyde oxime
Catalog # APOSPC10236-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:265.667 g/mol
Melting Pt:67…69 °C
Storage Temperature:Ambient
CAS Number:451485-70-4
Formula:C₁₃H₉ClFNO₂
MDL Number:MFCD02082405