Order Entry
Czech Republic
ContactUsLinkComponent
2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride
2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride
Catalog # APOSPC10205-500MG
Supplier:  Apollo Scientific
CAS Number:  
2-[(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}ethyl)amino]benzenaminium chloride
Catalog # APOSPC10205-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :367.197 g/mol
Melting Pt:175…185 °C
Storage Temperature:Ambient
CAS Number:317822-53-0
Formula:C₁₄H₁₅ClF₃N₄·Cl
MDL Number:MFCD00793064