Order Entry
Czech Republic
ContactUsLinkComponent
Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Irritant
Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Catalog # APOSPC1018-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Potassium perfluoro-4-methyl-3,6-dioxaoctane-1-sulphonate
Catalog # APOSPC1018-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :520.211 g/mol
Melting Pt:250 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:70755-50-9
Formula:C₇F₁₅O₅S·K
MDL Number:MFCD00155781