
1,4-Dihydro-4-oxo-6-(trifluoromethyl)quinoline-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:641993-21-7
:
Formula:C₁₁H₆F₃NO₃
MDL Number:MFCD00206334
:257.165 g/mol
Melting Pt:270…272 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together