
3,4,4,4-Tetrafluoro-3-(trifluoromethyl)butan-1-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:90999-87-4
:
Formula:C₅H₅F₇O
MDL Number:MFCD07368670
:214.081 g/mol
Boiling Pt:119…121 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together