

3,4,4-Trifluorobut-3-en-1-ol 97%
Catalog # APOSPC0990-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:126.077 g/mol
Boiling Pt:93 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07
CAS Number:97168-13-3
Formula:C₄H₅F₃O
MDL Number:MFCD07784254