
2,3-Difluoro-6-[(2,2-dimethylpropanoyl)amino]benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:874784-13-1
:
Formula:C₁₂H₁₃F₂NO₃
MDL Number:MFCD04035654
:257.233 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together