Order Entry
Czech Republic
ContactUsLinkComponent
(3S)-4,4,4-Trifluorobutane-1,3-diol 97%
Flammable
Irritant
(3S)-4,4,4-Trifluorobutane-1,3-diol 97%
Catalog # APOSPC0617-250MG
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
(3S)-4,4,4-Trifluorobutane-1,3-diol 97%
Catalog # APOSPC0617-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :144.092 g/mol
Boiling Pt:75…76 °C (5 mm Hg)
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:135154-88-0
Formula:C₄H₇F₃O₂
MDL Number:MFCD03093015